Product Details

SNP ID
rs150168776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:75732918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGACACACCTATGATATTGGAAT[G/T]ATCAGACAGGGATTTTAAAAAAAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR4273 PubMed Links

Gene Details

Gene
MIR4273
Gene Name
microRNA 4273
There are no transcripts associated with this gene.

Gene
ZNF717
Gene Name
zinc finger protein 717
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128223.2 Intron NP_001121695.1
NM_001290208.1 Intron NP_001277137.1
NM_001290209.1 Intron NP_001277138.1
NM_001290210.1 Intron NP_001277139.1
NM_001324026.1 Intron NP_001310955.1
NM_001324027.1 Intron NP_001310956.1
NM_001324028.1 Intron NP_001310957.1
XM_005264711.3 Intron XP_005264768.1
XM_006712912.3 Intron XP_006712975.1
XM_011533244.2 Intron XP_011531546.1
XM_011533245.2 Intron XP_011531547.1
XM_011533246.2 Intron XP_011531548.1
XM_011533248.2 Intron XP_011531550.1
XM_011533249.2 Intron XP_011531551.1
XM_011533250.2 Intron XP_011531552.1
XM_011533251.1 Intron XP_011531553.1
XM_017005486.1 Intron XP_016860975.1
XM_017005487.1 Intron XP_016860976.1

View Full Product Details