Product Details

SNP ID
rs28459556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:39052175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACGACCAGCTGGGGTAGCGGCA[A/T]GGGGCGAACGGGGCGGGGCGCGGCC
Phenotype
MIM: 604385 MIM: 612167
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SCN11A PubMed Links

Gene Details

Gene
SCN11A
Gene Name
sodium voltage-gated channel alpha subunit 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287223.1 Intron NP_001274152.1
NM_014139.2 Intron NP_054858.2
XM_011533321.2 Intron XP_011531623.1
XM_017005647.1 Intron XP_016861136.1
XM_017005648.1 Intron XP_016861137.1
XM_017005649.1 Intron XP_016861138.1
XM_017005650.1 Intron XP_016861139.1
XM_017005651.1 Intron XP_016861140.1
XM_017005652.1 Intron XP_016861141.1
XM_017005653.1 Intron XP_016861142.1
Gene
WDR48
Gene Name
WD repeat domain 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303402.1 Intron NP_001290331.1
NM_001303403.1 Intron NP_001290332.1
NM_020839.3 Intron NP_065890.1
XM_005265346.3 Intron XP_005265403.1
XM_011533971.1 Intron XP_011532273.1
XM_017006960.1 Intron XP_016862449.1

View Full Product Details