Product Details

SNP ID
rs28435231
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:171819360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTAGTTCATCACATTGCCCCATG[A/G]GAGACAGAGTGTGAAACACAGTCTT
Phenotype
MIM: 615138
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GALNTL6 PubMed Links

Gene Details

Gene
GALNTL6
Gene Name
polypeptide N-acetylgalactosaminyltransferase-like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034845.2 Intron NP_001030017.2
XM_011531993.2 Intron XP_011530295.1
XM_011531994.1 Intron XP_011530296.1
XM_011531995.1 Intron XP_011530297.1
XM_011531996.1 Intron XP_011530298.1
XM_011531997.1 Intron XP_011530299.1
XM_017008243.1 Intron XP_016863732.1
XM_017008244.1 Intron XP_016863733.1
Gene
LOC441052
Gene Name
uncharacterized LOC441052
There are no transcripts associated with this gene.

View Full Product Details