Product Details
- SNP ID
-
rs28702008
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:35888229 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAAGGATAATTTTCCTAACTGGAG[A/G]GAGATTAAAAAAGAAAAAAGAGAAA
- Phenotype
-
MIM: 606228
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AGO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs372107016] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGO1
- Gene Name
- argonaute 1, RISC catalytic component
View Full Product Details