Product Details

SNP ID
rs28373598
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:37960226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGATGGACTGAACATCTGTTGG[C/G]TATCCAGTGTTATAATCCTCTCTCC
Phenotype
MIM: 605596
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SF3A3 PubMed Links

Gene Details

Gene
SF3A3
Gene Name
splicing factor 3a subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320830.1 Intron NP_001307759.1
NM_006802.3 Intron NP_006793.1

View Full Product Details