Product Details

SNP ID
rs17885946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:102323961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGTAATGTATTATTTTCCTCCT[C/T]TGAGTTAGGTCTTGTGCTTTTTTTT
Phenotype
MIM: 601721
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BIRC3 PubMed Links

Gene Details

Gene
BIRC3
Gene Name
baculoviral IAP repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165.4 2247 UTR 5 NP_001156.1
NM_182962.2 2247 Intron NP_892007.1
XM_017017643.1 2247 UTR 5 XP_016873132.1

View Full Product Details