Product Details
- SNP ID
-
rs17881509
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:75402231 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAAAGCCATCTCCTGGGCAGCATG[C/T]GGCCCGTGGGTTGGGCAAGCTTGGT
- Phenotype
-
MIM: 600454
MIM: 600455
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RPS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141837939] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RPS3
- Gene Name
- ribosomal protein S3
- Gene
- SNORD15A
- Gene Name
- small nucleolar RNA, C/D box 15A
There are no transcripts associated with this gene.
- Gene
- SNORD15B
- Gene Name
- small nucleolar RNA, C/D box 15B
There are no transcripts associated with this gene.
View Full Product Details