Product Details

SNP ID
rs28817103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:940681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATATTTGCTTGTCAATTGTGCC[A/G]TATGTGCCACACCAATGTAAGATGT
Phenotype
MIM: 602052 MIM: 616660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAK PubMed Links

Gene Details

Gene
GAK
Gene Name
cyclin G associated kinase
There are no transcripts associated with this gene.

Gene
TMEM175
Gene Name
transmembrane protein 175
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297423.1 Intron NP_001284352.1
NM_001297424.1 Intron NP_001284353.1
NM_001297425.1 Intron NP_001284354.1
NM_001297426.1 Intron NP_001284355.1
NM_001297427.1 Intron NP_001284356.1
NM_001297428.1 Intron NP_001284357.1
NM_032326.3 Intron NP_115702.1
XM_005272301.2 Intron XP_005272358.1
XM_005272303.4 Intron XP_005272360.1
XM_005272304.1 Intron XP_005272361.1
XM_005272307.4 Intron XP_005272364.1
XM_011513576.1 Intron XP_011511878.1
XM_017008701.1 Intron XP_016864190.1
XM_017008702.1 Intron XP_016864191.1
XM_017008703.1 Intron XP_016864192.1
XM_017008704.1 Intron XP_016864193.1
XM_017008705.1 Intron XP_016864194.1

View Full Product Details