Product Details

SNP ID
rs61731350
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:152884461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAACACAGTAAATGCTGCCCAGC[A/G]AAAGGCAATCAATGCTGCCCACCAC
Phenotype
MIM: 601148
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMCP PubMed Links

Gene Details

Gene
SMCP
Gene Name
sperm mitochondria associated cysteine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030663.2 184 Silent Mutation GCA,GCG A13A NP_109588.2

View Full Product Details