Product Details

SNP ID
rs146206905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:120628497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCTTGAGTCCCCACTTTGTGAC[C/T]GCCAGACTTAAAGATGATTCAAACA
Phenotype
MIM: 607474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HGD PubMed Links

Gene Details

Gene
HGD
Gene Name
homogentisate 1,2-dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000187.3 1681 Silent Mutation GCA,GCG A407A NP_000178.2
XM_005247412.1 1681 Silent Mutation GCA,GCG A332A XP_005247469.1
XM_005247413.1 1681 Intron XP_005247470.1
XM_005247414.4 1681 Intron XP_005247471.1
XM_011512746.1 1681 Intron XP_011511048.1
XM_017006277.1 1681 Silent Mutation GCA,GCG A266A XP_016861766.1

View Full Product Details