Product Details

SNP ID
rs146078455
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:172889573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGTGCCTGCTCCCTAATGACTA[C/G]CTTTGCTGAACAGTTTGAAGTCGCT
Phenotype
MIM: 609856
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPATA16 PubMed Links

Gene Details

Gene
SPATA16
Gene Name
spermatogenesis associated 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031955.5 1891 Missense Mutation AGC,AGG S569R NP_114161.3
XM_006713778.2 1891 Intron XP_006713841.1
XM_017007308.1 1891 Intron XP_016862797.1

View Full Product Details