Product Details

SNP ID
rs116637064
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:19795338 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATGTAGTACGGGTTGAAGACTG[C/G]TTAAAAGTTTTCCCATATTCTATAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF506 PubMed Links

Gene Details

Gene
ZNF506
Gene Name
zinc finger protein 506
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099269.2 697 Missense Mutation AAC,AAG N183K NP_001092739.1
NM_001145404.1 697 Missense Mutation AAC,AAG N151K NP_001138876.1

View Full Product Details