Product Details
- SNP ID
-
rs61754182
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:20447669 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGGCGGGCCAGGAGGACTACGACCG[C/T]CTGCGGCCGCTCTCCTACCCGGACA
- Phenotype
-
MIM: 165370
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RHOB
PubMed Links
Gene Details
- Gene
- RHOB
- Gene Name
- ras homolog family member B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004040.3 |
599 |
Silent Mutation |
CGC,CGT |
R68R |
NP_004031.1 |
View Full Product Details