Product Details

SNP ID
rs41265059
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:20500049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCAATCTGTTGCCCGGGGCCCC[A/G]ACCATCTGGGCAGAAGCGGATGTGC
Phenotype
MIM: 612037
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MUL1 PubMed Links

Gene Details

Gene
MUL1
Gene Name
mitochondrial E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024544.2 2540 UTR 3 NP_078820.2
XM_011542137.2 2540 UTR 3 XP_011540439.1

View Full Product Details