Product Details

SNP ID
rs41285429
Assay Type
Functionally Tested
NCBI dbSNP Submissions
14
Location
Chr.1:26320292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAATGCCATAATCCACCTCTTCT[A/G]CTTCAGTTGAGGTGACACGTCTCAG
Phenotype
MIM: 114280 MIM: 609151
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
AIM1L PubMed Links

Gene Details

Gene
AIM1L
Gene Name
absent in melanoma 1-like
There are no transcripts associated with this gene.

Gene
CD52
Gene Name
CD52 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001803.2 274 Missense Mutation TAC,TGC Y59C NP_001794.2
Gene
UBXN11
Gene Name
UBX domain protein 11
There are no transcripts associated with this gene.

View Full Product Details