Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_013261.3 | 5333 | UTR 3 | NP_037393.1 | ||
XM_005248131.4 | 5333 | UTR 3 | XP_005248188.1 | ||
XM_005248132.1 | 5333 | UTR 3 | XP_005248189.1 | ||
XM_005248134.4 | 5333 | UTR 3 | XP_005248191.1 | ||
XM_011513765.2 | 5333 | UTR 3 | XP_011512067.1 | ||
XM_011513766.1 | 5333 | UTR 3 | XP_011512068.1 | ||
XM_011513767.2 | 5333 | UTR 3 | XP_011512069.1 | ||
XM_011513768.1 | 5333 | UTR 3 | XP_011512070.1 | ||
XM_011513769.2 | 5333 | Intron | XP_011512071.1 | ||
XM_011513770.2 | 5333 | UTR 3 | XP_011512072.1 | ||
XM_011513771.1 | 5333 | UTR 3 | XP_011512073.1 | ||
XM_017007664.1 | 5333 | UTR 3 | XP_016863153.1 |