Product Details

SNP ID
rs45535333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:113860153 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTGAGGCGCCCGCTGCCGAAGG[C/T]GGCAAGGACGAGGCCGCCGGGGGCG
Phenotype
MIM: 177061
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC01268 PubMed Links

Gene Details

Gene
LINC01268
Gene Name
long intergenic non-protein coding RNA 1268
There are no transcripts associated with this gene.

Gene
MARCKS
Gene Name
myristoylated alanine rich protein kinase C substrate
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002356.6 984 Silent Mutation GGC,GGT G191G NP_002347.5

View Full Product Details