Product Details

SNP ID
rs45483993
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:46799632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGGCAATCCATTGAAAGCTCAA[C/T]CAGCGTCCGTGTTGTTGTGTGTAGT
Phenotype
MIM: 124075
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYP4B1 PubMed Links

Gene Details

Gene
CYP4B1
Gene Name
cytochrome P450 family 4 subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000779.3 Intron NP_000770.2
NM_001099772.1 Intron NP_001093242.1
NM_001319161.1 Intron NP_001306090.1
NM_001319162.1 Intron NP_001306091.1
NM_001319163.1 Intron NP_001306092.1
XM_011540832.1 Intron XP_011539134.1
XM_011540833.1 Intron XP_011539135.1
XM_017000466.1 Intron XP_016855955.1

View Full Product Details