Product Details

SNP ID
rs61758714
Assay Type
Functionally tested
NCBI dbSNP Submissions
36
Location
Chr.1:202135488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAATCAGCTCCTTCTCATCCAA[A/T]GCTCCCGGAAGGTCTCGCTTGTTAC
Phenotype
MIM: 616597
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARL8A PubMed Links

Gene Details

Gene
ARL8A
Gene Name
ADP ribosylation factor like GTPase 8A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256129.1 582 Intron NP_001243058.1
NM_138795.3 582 Silent Mutation GCA,GCT A137A NP_620150.1
Gene
GPR37L1
Gene Name
G protein-coupled receptor 37 like 1
There are no transcripts associated with this gene.

View Full Product Details