Product Details

SNP ID
rs62187656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161631189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAGGTACAGAGTTTCTTTTTGC[A/G]GTGATGGAAATGTTTTGAAATTACG
Phenotype
MIM: 605556
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC4A10 PubMed Links

Gene Details

Gene
SLC4A10
Gene Name
solute carrier family 4 member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178015.1 Intron NP_001171486.1
NM_001178016.1 Intron NP_001171487.1
NM_022058.3 Intron NP_071341.2
XM_005246694.4 Intron XP_005246751.1
XM_005246695.4 Intron XP_005246752.1
XM_011511508.2 Intron XP_011509810.1
XM_011511509.2 Intron XP_011509811.1
XM_011511510.2 Intron XP_011509812.1
XM_011511511.2 Intron XP_011509813.1
XM_011511512.2 Intron XP_011509814.1
XM_011511513.2 Intron XP_011509815.1
XM_017004541.1 Intron XP_016860030.1
XM_017004542.1 Intron XP_016860031.1
XM_017004543.1 Intron XP_016860032.1
XM_017004544.1 Intron XP_016860033.1
XM_017004545.1 Intron XP_016860034.1
XM_017004546.1 Intron XP_016860035.1
XM_017004547.1 Intron XP_016860036.1
XM_017004548.1 Intron XP_016860037.1
XM_017004549.1 Intron XP_016860038.1
XM_017004550.1 Intron XP_016860039.1
XM_017004551.1 Intron XP_016860040.1
XM_017004552.1 Intron XP_016860041.1
XM_017004553.1 Intron XP_016860042.1
XM_017004554.1 Intron XP_016860043.1

View Full Product Details