Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001305008.1 | 3966 | UTR 3 | NP_001291937.1 | ||
NM_001305009.1 | 3966 | UTR 3 | NP_001291938.1 | ||
NM_001305010.1 | 3966 | UTR 3 | NP_001291939.1 | ||
NM_001305011.1 | 3966 | UTR 3 | NP_001291940.1 | ||
NM_030650.2 | 3966 | UTR 3 | NP_085153.1 | ||
XM_005246878.3 | 3966 | UTR 3 | XP_005246935.1 | ||
XM_005246879.4 | 3966 | UTR 3 | XP_005246936.1 | ||
XM_005246880.3 | 3966 | UTR 3 | XP_005246937.1 | ||
XM_006712783.2 | 3966 | UTR 3 | XP_006712846.1 | ||
XM_011511945.2 | 3966 | UTR 3 | XP_011510247.1 | ||
XM_017005055.1 | 3966 | UTR 3 | XP_016860544.1 |