Product Details

SNP ID
hCV88489366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:119500853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAAATTTATCATAACCGGTTTG[A/G]TGCTGTGGTATGTACTGGTGATCTA
Phenotype
MIM: 615618 MIM: 615534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POGLUT1 PubMed Links

Gene Details

Gene
POGLUT1
Gene Name
protein O-glucosyltransferase 1
There are no transcripts associated with this gene.

Gene
TIMMDC1
Gene Name
translocase of inner mitochondrial membrane domain containing 1
There are no transcripts associated with this gene.

View Full Product Details