Product Details

SNP ID
rs58850362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136873990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTATACCTTTTTGTGGATGGCT[A/G]CTTTTGCTTAACATTGTGTTTGTGA
Phenotype
MIM: 600508
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCK1 PubMed Links

Gene Details

Gene
NCK1
Gene Name
NCK adaptor protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190796.2 Intron NP_001177725.1
NM_001291999.1 Intron NP_001278928.1
NM_006153.5 Intron NP_006144.1

View Full Product Details