Product Details
- SNP ID
-
rs56911212
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:88762628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAACTTTATTGATTTTTTTCCTTT[A/G]GTATAAATGTTAAACTCAGAGATCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF804B
PubMed Links
Gene Details
- Gene
- ZNF804B
- Gene Name
- zinc finger protein 804B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181646.3 |
|
Intron |
|
|
NP_857597.1 |
View Full Product Details