Product Details

SNP ID
rs61749953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:108571730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTGTTTTAGGATATTAGAAATG[A/G]CTACTCCCCAGTCAATTTTCATCTT
Phenotype
MIM: 602634 MIM: 612534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJB9 PubMed Links

Gene Details

Gene
DNAJB9
Gene Name
DnaJ heat shock protein family (Hsp40) member B9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012328.2 373 Missense Mutation ACT,GCT T2A NP_036460.1
Gene
THAP5
Gene Name
THAP domain containing 5
There are no transcripts associated with this gene.

View Full Product Details