Product Details

SNP ID
rs62506083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:51820568 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATTACCCACAAATACGTAAGTG[G/T]TAATTCTCTGTTTAACTCTCTTTCT
Phenotype
MIM: 615904
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCMTD1 PubMed Links

Gene Details

Gene
PCMTD1
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286782.1 704 Missense Mutation AAC,ACC N210T NP_001273711.1
NM_001286783.1 704 Missense Mutation AAC,ACC N110T NP_001273712.1
NM_052937.3 704 Missense Mutation AAC,ACC N286T NP_443169.2
XM_005251146.2 704 Intron XP_005251203.1
XM_011517444.2 704 Missense Mutation AAC,ACC N110T XP_011515746.1
XM_017013017.1 704 Missense Mutation AAC,ACC N110T XP_016868506.1
Gene
PXDNL
Gene Name
peroxidasin like
There are no transcripts associated with this gene.

View Full Product Details