Product Details

SNP ID
rs61931389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31960551 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATTGTTTTGCAGTGATACTACA[A/G]TAGCGGACCTAGAGGAGCGCACATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA1551 PubMed Links

Gene Details

Gene
KIAA1551
Gene Name
KIAA1551
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018169.3 Intron NP_060639.3
XM_005253405.2 Intron XP_005253462.1
XM_006719099.2 Intron XP_006719162.1
XM_011520722.1 Intron XP_011519024.1
XM_017019550.1 Intron XP_016875039.1

View Full Product Details