Product Details
- SNP ID
-
hCV89142940
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:14882281 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACAGGCTTAGAGCGTTCTCGGATC[A/C]TAGAAAGTGGAAGAAGAGGCCAAAA
- Phenotype
-
MIM: 154870
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MGP
PubMed Links
Gene Details
- Gene
- MGP
- Gene Name
- matrix Gla protein
There are no transcripts associated with this gene.
View Full Product Details