Product Details

SNP ID
rs60239792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122475428 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCACTGACCATCATCACCTACA[C/T]GTTCCACAGATACAACCTGTCCAAA
Phenotype
MIM: 616381
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZCCHC8 PubMed Links

Gene Details

Gene
ZCCHC8
Gene Name
zinc finger CCHC-type containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017612.4 Intron NP_060082.2
XM_006719491.2 Intron XP_006719554.1
XM_011538555.2 Intron XP_011536857.1
XM_017019606.1 Intron XP_016875095.1

View Full Product Details