Product Details

SNP ID
rs56289107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40334777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTGGAGAGGGAGGCCAGGCCCTG[C/G]GGGGGTGACGGGGAGCCAGGAGAGC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C15orf52 PubMed Links

Gene Details

Gene
C15orf52
Gene Name
chromosome 15 open reading frame 52
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207380.2 2002 UTR 3 NP_997263.2
Gene
INAFM2
Gene Name
InaF motif containing 2
There are no transcripts associated with this gene.

View Full Product Details