Product Details

SNP ID
hCV89281504
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69316021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCTTTCCAGAAAGCCATTGATC[A/T]GGTGACGAAAGCCACAGAGGAGGAC
Phenotype
MIM: 600027 MIM: 609982
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SNTB2 PubMed Links

Gene Details

Gene
SNTB2
Gene Name
syntrophin beta 2
There are no transcripts associated with this gene.

Gene
VPS4A
Gene Name
vacuolar protein sorting 4 homolog A
There are no transcripts associated with this gene.

View Full Product Details