Product Details

SNP ID
rs56158152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84700541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGGTTGCTGCCTCTGCCTGGAGC[G/T]AGCCTCAGAGATTTGGGGGTGCCCT
Phenotype
MIM: 609818
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
USP10 PubMed Links

Gene Details

Gene
USP10
Gene Name
ubiquitin specific peptidase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272075.1 Intron NP_001259004.1
NM_005153.2 Intron NP_005144.2
XM_006721332.1 Intron XP_006721395.1
XM_011523440.1 Intron XP_011521742.1
XM_011523441.1 Intron XP_011521743.1
XM_011523443.1 Intron XP_011521745.1
XM_017023863.1 Intron XP_016879352.1
XM_017023864.1 Intron XP_016879353.1
XM_017023865.1 Intron XP_016879354.1
XM_017023866.1 Intron XP_016879355.1
XM_017023867.1 Intron XP_016879356.1
XM_017023868.1 Intron XP_016879357.1
XM_017023869.1 Intron XP_016879358.1

View Full Product Details