Product Details

SNP ID
rs60688184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73193327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGCCGTGAAGGCCACCGACCAGTA[C/G]TGCGCCCGCCTCCGCCAGGCCGGCT
Phenotype
MIM: 606973
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
COG1 PubMed Links

Gene Details

Gene
COG1
Gene Name
component of oligomeric golgi complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018714.2 294 Silent Mutation TAC,TAG Y86* NP_061184.1

View Full Product Details