Product Details

SNP ID
rs62058111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10392865 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTTCTGGATCTGCTTCTTCCCA[A/C]CCTTCAGCGCCAGCTGCTCGGCCTC
Phenotype
MIM: 160741
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MYH8 PubMed Links

Gene Details

Gene
MYH8
Gene Name
myosin heavy chain 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002472.2 5524 Missense Mutation GGT,GTT G1810V NP_002463.2
Gene
MYHAS
Gene Name
myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.

View Full Product Details