Product Details
- SNP ID
-
hCV89377744
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:27760664 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGGAGCCGTTGCTGCCAGAAACTG[A/C]GGAAGGGCGCGATGCCTGTGCCAGG
- Phenotype
-
MIM: 163730
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NOS2
PubMed Links
Gene Details
- Gene
- NOS2
- Gene Name
- nitric oxide synthase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000625.4 |
3233 |
Missense Mutation |
CGC,CTC |
R990L |
NP_000616.3 |
View Full Product Details