Product Details

SNP ID
rs59566682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:2919241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTCTGCTCCATAGGAGCTCCA[A/G]GACCCCTCTTTGACGCTGCTTCCAC
Phenotype
MIM: 608928 MIM: 605519
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EMILIN2 PubMed Links

Gene Details

Gene
EMILIN2
Gene Name
elastin microfibril interfacer 2
There are no transcripts associated with this gene.

Gene
LPIN2
Gene Name
lipin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014646.2 3842 UTR 3 NP_055461.1
XM_005258177.4 3842 UTR 3 XP_005258234.1
XM_005258178.3 3842 UTR 3 XP_005258235.1
XM_005258179.4 3842 UTR 3 XP_005258236.1
XM_017026098.1 3842 UTR 3 XP_016881587.1
XM_017026099.1 3842 UTR 3 XP_016881588.1

View Full Product Details