Product Details

SNP ID
hCV89429288
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44907771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTTCCACACAGGATGCCAGGCC[A/G]AGGTGGAGCAAGCGGTGGAGACAGA
Phenotype
MIM: 107710 MIM: 107741 MIM: 608061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APOC1 PubMed Links

Gene Details

Gene
APOC1
Gene Name
apolipoprotein C1
There are no transcripts associated with this gene.

Gene
APOE
Gene Name
apolipoprotein E
There are no transcripts associated with this gene.

Gene
TOMM40
Gene Name
translocase of outer mitochondrial membrane 40
There are no transcripts associated with this gene.

View Full Product Details