Product Details

SNP ID
rs56107738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:46307488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTCAGTGTGCTTTTTAGCACCT[A/G]ACAGACTATCACTAAACTTCATCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C21orf58 PubMed Links

Gene Details

Gene
C21orf58
Gene Name
chromosome 21 open reading frame 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286462.1 Intron NP_001273391.1
NM_001286463.1 Intron NP_001273392.1
NM_001286476.1 Intron NP_001273405.1
NM_001286477.1 Intron NP_001273406.1
NM_058180.4 Intron NP_478060.2
XM_005261148.4 Intron XP_005261205.1
XM_006724018.3 Intron XP_006724081.1
XM_011529619.2 Intron XP_011527921.1
XM_011529620.2 Intron XP_011527922.1
XM_011529621.2 Intron XP_011527923.1
XM_011529623.2 Intron XP_011527925.1
XM_011529624.2 Intron XP_011527926.1
XM_011529625.2 Intron XP_011527927.1
XM_011529626.2 Intron XP_011527928.1
XM_011529627.2 Intron XP_011527929.1
XM_011529628.1 Intron XP_011527930.1
XM_017028388.1 Intron XP_016883877.1
XM_017028389.1 Intron XP_016883878.1
XM_017028390.1 Intron XP_016883879.1
XM_017028391.1 Intron XP_016883880.1
XM_017028392.1 Intron XP_016883881.1
XM_017028393.1 Intron XP_016883882.1
Gene
YBEY
Gene Name
ybeY metallopeptidase (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006114.2 Intron NP_001006114.1
NM_001314022.1 Intron NP_001300951.1
NM_001314023.1 Intron NP_001300952.1
NM_001314024.1 Intron NP_001300953.1
NM_001314025.1 Intron NP_001300954.1
NM_001314026.1 Intron NP_001300955.1
NM_058181.2 Intron NP_478061.1
XM_005261156.3 Intron XP_005261213.2
XM_011529629.2 Intron XP_011527931.1
XM_011529630.2 Intron XP_011527932.1
XM_011529631.2 Intron XP_011527933.1
XM_011529633.2 Intron XP_011527935.1
XM_011529634.2 Intron XP_011527936.2
XM_011529635.2 Intron XP_011527937.1
XM_017028394.1 Intron XP_016883883.1
XM_017028395.1 Intron XP_016883884.1
XM_017028396.1 Intron XP_016883885.1

View Full Product Details