Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001006114.2 | 1648 | Intron | NP_001006114.1 | ||
NM_001314022.1 | 1648 | Intron | NP_001300951.1 | ||
NM_001314023.1 | 1648 | Intron | NP_001300952.1 | ||
NM_001314024.1 | 1648 | Intron | NP_001300953.1 | ||
NM_001314025.1 | 1648 | Intron | NP_001300954.1 | ||
NM_001314026.1 | 1648 | Intron | NP_001300955.1 | ||
NM_058181.2 | 1648 | Intron | NP_478061.1 | ||
XM_005261156.3 | 1648 | Intron | XP_005261213.2 | ||
XM_011529629.2 | 1648 | Intron | XP_011527931.1 | ||
XM_011529630.2 | 1648 | Intron | XP_011527932.1 | ||
XM_011529631.2 | 1648 | Intron | XP_011527933.1 | ||
XM_011529633.2 | 1648 | UTR 3 | XP_011527935.1 | ||
XM_011529634.2 | 1648 | Intron | XP_011527936.2 | ||
XM_011529635.2 | 1648 | Intron | XP_011527937.1 | ||
XM_017028394.1 | 1648 | UTR 3 | XP_016883883.1 | ||
XM_017028395.1 | 1648 | Intron | XP_016883884.1 | ||
XM_017028396.1 | 1648 | Intron | XP_016883885.1 |