Product Details

SNP ID
rs58368178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41375051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCACCTCTGTCCCCTCCAGCA[C/T]GTCTTTGACTGCAGAAACCGTGGCT
Phenotype
MIM: 188595
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100996598 PubMed Links

Gene Details

Gene
LOC100996598
Gene Name
uncharacterized LOC100996598
There are no transcripts associated with this gene.

Gene
TEF
Gene Name
TEF, PAR bZIP transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145398.2 Intron NP_001138870.1
NM_003216.3 Intron NP_003207.1

View Full Product Details