Product Details
- SNP ID
-
hCV89667257
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:87977078 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTAAGGTTAAACAGGCTGATTCTG[A/G]AAGTTCTGAGGAAAAGCAGGTAAGC
- Phenotype
-
MIM: 166490
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SPP1
PubMed Links
Gene Details
- Gene
- SPP1
- Gene Name
- secreted phosphoprotein 1
There are no transcripts associated with this gene.
View Full Product Details