Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001080473.1 | Intron | NP_001073942.1 | |||
XM_005264301.4 | Intron | XP_005264358.1 | |||
XM_017004063.1 | Intron | XP_016859552.1 | |||
XM_017004064.1 | Intron | XP_016859553.1 | |||
XM_017004065.1 | Intron | XP_016859554.1 | |||
XM_017004066.1 | Intron | XP_016859555.1 | |||
XM_017004067.1 | Intron | XP_016859556.1 | |||
XM_017004068.1 | Intron | XP_016859557.1 | |||
XM_017004069.1 | Intron | XP_016859558.1 | |||
XM_017004070.1 | Intron | XP_016859559.1 |