Product Details

SNP ID
rs56386759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9505053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCTGTGATGTGAACATGTCAC[A/G]TGTATTTATCCGGCCTCCTGTCCAT
Phenotype
MIM: 610240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LHFPL4 PubMed Links

Gene Details

Gene
LHFPL4
Gene Name
lipoma HMGIC fusion partner-like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198560.2 Intron NP_940962.1
XM_017006359.1 Intron XP_016861848.1
XM_017006360.1 Intron XP_016861849.1

View Full Product Details