Product Details

SNP ID
rs55928815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:31207094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTGAGTGGTACTGAAAGGGGTA[C/T]TTACTTTATCTGTATGTCTGTTTAT
Phenotype
MIM: 610308
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GLCT PubMed Links

Gene Details

Gene
B3GLCT
Gene Name
beta 3-glucosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_194318.3 Intron NP_919299.3
XM_006719768.3 Intron XP_006719831.1
XM_011534936.1 Intron XP_011533238.1
XM_011534938.2 Intron XP_011533240.1
XM_017020395.1 Intron XP_016875884.1

View Full Product Details