Product Details

SNP ID
rs57387056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77769594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACATCTGGAGTCAGACAGACCTGA[A/G]TTCAAGTCGTGCCTTCCTCCCCACT
Phenotype
MIM: 603055
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf178 PubMed Links

Gene Details

Gene
C14orf178
Gene Name
chromosome 14 open reading frame 178
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173978.1 484 Silent Mutation GAA,GAG E65E NP_001167449.1
NM_174943.3 484 Silent Mutation GAA,GAG E95E NP_777603.1
Gene
SNW1
Gene Name
SNW domain containing 1
There are no transcripts associated with this gene.

View Full Product Details