Product Details

SNP ID
rs56117810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:62262466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTAAAATAGAAATCTGTAAGTCT[A/G]TAAGTTAAAATATAAGTGTTATTGT
Phenotype
MIM: 604731
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6125 PubMed Links

Gene Details

Gene
MIR6125
Gene Name
microRNA 6125
There are no transcripts associated with this gene.

Gene
USP15
Gene Name
ubiquitin specific peptidase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252078.1 Intron NP_001239007.1
NM_001252079.1 Intron NP_001239008.1
NM_006313.2 Intron NP_006304.1
XM_005269261.2 Intron XP_005269318.1
XM_006719718.3 Intron XP_006719781.1
XM_017020284.1 Intron XP_016875773.1
XM_017020285.1 Intron XP_016875774.1

View Full Product Details