Product Details

SNP ID
rs55946540
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:86931106 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAATGATGTTGTAGTATGTGAC[A/G]TGAAATATAGTAGGTTTTGAGAGTT
Phenotype
MIM: 604844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HS2ST1 PubMed Links

Gene Details

Gene
HS2ST1
Gene Name
heparan sulfate 2-O-sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134492.1 Intron NP_001127964.1
NM_012262.3 Intron NP_036394.1

View Full Product Details