Product Details

SNP ID
rs60079577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55638555 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTTCCTCAATTACCTCTCACT[C/G]TCCGACCTTTGCTACACATCCACAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR4P4 PubMed Links

Gene Details

Gene
OR4P4
Gene Name
olfactory receptor family 4 subfamily P member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004124.2 198 Silent Mutation CTC,CTG L66L NP_001004124.1

View Full Product Details