Product Details

SNP ID
rs61876245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:319713 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTTTATTGAATGCCATTGTAG[A/G]AAAGCGTGTGAGGATAAAGGGCTGA
Phenotype
MIM: 604456 MIM: 605579
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFITM1 PubMed Links

Gene Details

Gene
IFITM1
Gene Name
interferon induced transmembrane protein 1
There are no transcripts associated with this gene.

Gene
IFITM3
Gene Name
interferon induced transmembrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021034.2 Intron NP_066362.2

View Full Product Details