Product Details

SNP ID
rs74116988
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:157133446 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCACAAACAAAAAACCACACAA[C/T]AACAAAGTCTTTTGAGAGACTGCTT
Phenotype
MIM: 164873
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYCSP52 PubMed Links

Gene Details

Gene
CYCSP52
Gene Name
cytochrome c, somatic pseudogene 52
There are no transcripts associated with this gene.

Gene
ETV3
Gene Name
ETS variant 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145312.1 1153 Intron NP_001138784.1
NM_005240.1 1153 UTR 3 NP_005231.1
XM_006711210.2 1153 Intron XP_006711273.1

View Full Product Details